Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCF23 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TCF23 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP191533
![]() |
Novus Biologicals
NBP191533 |
100 μL |
Each for $487.50
|
|
|||||
NBP19153320
![]() |
Novus Biologicals
NBP19153320UL |
20 μL | N/A | N/A | N/A | ||||
Description
TCF23 Polyclonal specifically detects TCF23 in Mouse samples. It is validated for Western Blot.Specifications
TCF23 | |
Polyclonal | |
Purified | |
RUO | |
NP_444315 | |
150921 | |
Synthetic peptide directed towards the C terminal of mouse TCF23. Peptide sequence PMRSRLYAGGLGCSDLDSTTAITTGQRCKDAELGSQDSVAAESLLTSPAF. | |
Primary | |
23 kDa |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
BHLHA24, class A basic helix-loop-helix protein 24, class II basic helix-loop-helix protein TCF23, TCF-23, transcription factor 23 | |
TCF23 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title