Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Islet-2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP19153520UL

 View more versions of this product

Catalog No. NBP19153520


Only null left
Add to Cart

Description

Description

Islet-2 Polyclonal specifically detects Islet-2 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Islet-2
Polyclonal
Western Blot 1:1000
NP_081673
ISL2
Synthetic peptide directed towards the C terminal of mouse ISL2. Peptide sequence VIRVWFQNKRCKDKKKSILMKQLQQQQHSDKASFQGLTGTPLVAGSPIGH.
Protein A purified
RUO
64843
Store at -20C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
FLJ10160, insulin gene enhancer protein ISL-2, ISL LIM homeobox 2, ISL2 transcription factor, LIM/homeodomain, (islet-2), islet-2
Rabbit
39 kDa
20 μL
Primary
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.