Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | TTC19 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
TTC19 Polyclonal specifically detects TTC19 in Mouse samples. It is validated for Western Blot.Specifications
| TTC19 | |
| Polyclonal | |
| Purified | |
| RUO | |
| MGC138312, MGC19520, tetratricopeptide repeat domain 19,2010204O13Rik, tetratricopeptide repeat protein 19, mitochondrial, TPR repeat protein 19 | |
| TTC19 | |
| IgG | |
| Protein A purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| NP_082636 | |
| 54902 | |
| Synthetic peptide directed towards the N terminal of mouse TTC19. Peptide sequence RTRGAPARGHGVLPLLAALAWFSRPAATAEQPGEDASDEAEAEIIQLLKQ. | |
| Primary | |
| 39 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title