Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRSF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP182399
Description
NRSF Polyclonal specifically detects NRSF in Mouse samples. It is validated for Western Blot.Specifications
| NRSF | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5978 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence | |
| NP_035393 | |
| REST | |
| Synthetic peptide towards Rest. Peptide sequence ANMGMALTNDMYDLHELSKAELAAPQLIMLANVALTGEASGSCCDYLVGE. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Rabbit: 100%; Rat: 100%; Chicken: 92%; Mouse: 92%; Bovine: 85%; Pig: 85%; Xenopus: 84%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction