Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRSF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18239920UL
Description
NRSF Polyclonal specifically detects NRSF in Mouse samples. It is validated for Western Blot.Specifications
| NRSF | |
| Polyclonal | |
| Western Blot 1:1000 | |
| NP_035393 | |
| REST | |
| Synthetic peptide towards Rest. Peptide sequence ANMGMALTNDMYDLHELSKAELAAPQLIMLANVALTGEASGSCCDYLVGE. | |
| 20 μL | |
| Primary | |
| Mouse | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS & 2% Sucrose. with No Preservative | |
| Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5978 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction