Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mohawk homeobox Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Mohawk homeobox |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18240820
![]() |
Novus Biologicals
NBP18240820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP182408
![]() |
Novus Biologicals
NBP182408 |
100 μL |
Each for $487.50
|
|
|||||
Description
Mohawk homeobox Polyclonal specifically detects Mohawk homeobox in Mouse samples. It is validated for Western Blot.Specifications
Mohawk homeobox | |
Polyclonal | |
Rabbit | |
NP_808263 | |
283078 | |
Synthetic peptide towards mohawk homebox. Peptide sequence GDSAANRRGPSKDDTYWKEINAAMALTNLAQGKDEVQGTTTSCIIQKSSH. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
C10orf48, homeobox protein Mohawk, IFRX, Iroquois family related homeodomain protein, iroquois homeobox protein-like 1, IRXL1chromosome 10 open reading frame 48, MGC39616, mohawk homeobox | |
MKX | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title