Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFSD12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | MFSD12 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1915920
![]() |
Novus Biologicals
NBP19159120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191591
![]() |
Novus Biologicals
NBP191591 |
100 μL |
Each for $487.50
|
|
|||||
Description
MFSD12 Polyclonal specifically detects MFSD12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MFSD12 | |
Polyclonal | |
Rabbit | |
Human | |
C19orf28, chromosome 19 open reading frame 28, hypothetical protein LOC126321, major facilitator superfamily domain containing 12, MGC20700, PP3501 | |
MFSD12 | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
NP_001036145 | |
126321 | |
Synthetic peptide directed towards the middle region of human C19orf28. Peptide sequence VELTALRYAFTVVANITVYGAAWLLLHLQGSSRVEPTQDISISDQLGGQD. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title