Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

kinectin Antibody, Novus Biologicals™
SDP

Catalog No. NBP19159220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19159220 20 μL
NBP191592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP19159220 Supplier Novus Biologicals Supplier No. NBP19159220UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

kinectin Polyclonal specifically detects kinectin in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen kinectin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_001072990
Gene Alias kinectin, kinectin 1 (kinesin receptor), MGC133337
Gene Symbols KTN1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KTN1. Peptide sequence EELLKVISEREKEISGLWNELDSLKDAVEHQRKKNNDLREKNWEAMEALA.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3895
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.