Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RADIL Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP19161520UL

 View more versions of this product

Catalog No. NBP19161520


Only null left
Add to Cart

Description

Description

RADIL Polyclonal specifically detects RADIL in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

RADIL
Polyclonal
Western Blot 1:1000
NP_848817
RADIL
Synthetic peptide directed towards the C terminal of mouse D930005D10RIK. Peptide sequence PEPSGLCGRAVLDGQRNGPGGLPGAVLEGDAIQDAEPPAEASSPSSSAED.
20 μL
Primary
Mouse
Purified
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
FLJ10324, FLJ60975, KIAA1849MGC161589, Rap GTPase interactor, Ras association and DIL domains, ras-associating and dilute domain-containing protein, RASIP2
Rabbit
Protein A purified
RUO
55698
Store at -20C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.