Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RADIL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | RADIL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19161520
![]() |
Novus Biologicals
NBP19161520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191615
![]() |
Novus Biologicals
NBP191615 |
100 μL |
Each for $487.50
|
|
|||||
Description
RADIL Polyclonal specifically detects RADIL in Mouse samples. It is validated for Western Blot.Specifications
RADIL | |
Polyclonal | |
Purified | |
RUO | |
NP_848817 | |
55698 | |
Synthetic peptide directed towards the C terminal of mouse D930005D10RIK. Peptide sequence PEPSGLCGRAVLDGQRNGPGGLPGAVLEGDAIQDAEPPAEASSPSSSAED. | |
Primary |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
FLJ10324, FLJ60975, KIAA1849MGC161589, Rap GTPase interactor, Ras association and DIL domains, ras-associating and dilute domain-containing protein, RASIP2 | |
RADIL | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title