Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCORL Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP19161820UL

 View more versions of this product

Catalog No. NBP19161820


Only null left
Add to Cart

Description

Description

LCORL Polyclonal specifically detects LCORL in Human, Bovine samples. It is validated for Western Blot.
Specifications

Specifications

LCORL
Polyclonal
Western Blot 1.25 ug/ml
NP_742165
LCORL
Synthetic peptide directed towards the C terminal of human A830039H10RIK. Peptide sequence EIMEEAIAMVMSGKMSVSKAQGIYGVPHSTLEYKVKERSGTLKTPPKKKL.
20 μL
Primary
Human, Mouse
Purified
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
FLJ30696, LCoR-like protein, ligand dependent nuclear receptor corepressor-like, ligand-dependent nuclear receptor corepressor-like protein, MLR1MBLK1-related protein, transcription factor MLR1
Rabbit
Protein A purified
RUO
254251
Store at -20C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.