Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCORL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | LCORL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19161820
![]() |
Novus Biologicals
NBP19161820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191618
![]() |
Novus Biologicals
NBP191618 |
100 μL |
Each for $487.50
|
|
|||||
Description
LCORL Polyclonal specifically detects LCORL in Human, Bovine samples. It is validated for Western Blot.Specifications
LCORL | |
Polyclonal | |
Purified | |
RUO | |
FLJ30696, LCoR-like protein, ligand dependent nuclear receptor corepressor-like, ligand-dependent nuclear receptor corepressor-like protein, MLR1MBLK1-related protein, transcription factor MLR1 | |
LCORL | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
NP_742165 | |
254251 | |
Synthetic peptide directed towards the C terminal of human A830039H10RIK. Peptide sequence EIMEEAIAMVMSGKMSVSKAQGIYGVPHSTLEYKVKERSGTLKTPPKKKL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title