Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | YL1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
YL1 Polyclonal specifically detects YL1 in Mouse samples. It is validated for Western Blot.Specifications
| YL1 | |
| Polyclonal | |
| Rabbit | |
| NP_033362 | |
| 6944 | |
| Synthetic peptide towards YL1. Peptide sequence AGGRAPRKTAGNRLSGLLEAEEEDEFYQTTYGGFTEESGDDEYQGDQSDT. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Protein YL-1, Swc2, TCFL1, Transcription factor-like 1, transcription factor-like 1 (YL-1), vacuolar protein sorting 72 (yeast), vacuolar protein sorting 72 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 72 homolog, YL-1, YL1CFL1 | |
| VPS72 | |
| IgG | |
| 41 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title