Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIP5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | RIP5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19133220
![]() |
Novus Biologicals
NBP19133220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191332
![]() |
Novus Biologicals
NBP191332 |
100 μL |
Each for $487.50
|
|
|||||
Description
RIP5 Polyclonal specifically detects RIP5 in Human samples. It is validated for Western Blot.Specifications
RIP5 | |
Polyclonal | |
Purified | |
RUO | |
C20orf24, chromosome 20 open reading frame 24, PNAS-11 | |
Synthetic peptide directed towards the N terminal of human C20orf24. Peptide sequence MSGGRRKEEPPQPQLANGALKVSVWSKVLRSDAAWEDKDEFLDVIYWFRQ. | |
Primary |
Western Blot | |
Unconjugated | |
Rabbit | |
NP_061328 | |
55969 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title