Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBTD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19137520UL
Description
MBTD1 Polyclonal specifically detects MBTD1 in Mouse samples. It is validated for Western Blot.Specifications
MBTD1 | |
Polyclonal | |
Western Blot 1:1000 | |
NP_598773 | |
MBTD1 | |
The specific Immunogen is proprietary information. Peptide sequence LVPPRTVQHKYTNWKAFLVKRLTGAKTLPPDFSQKVSESMQYPFKPCMRV. | |
20 μL | |
Primary | |
Mouse | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
mbt domain containing 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
54799 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction