Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP9 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB184822 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184822 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184822 Supplier Novus Biologicals Supplier No. NBP282080B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NLRP9 Polyclonal antibody specifically detects NLRP9 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NLRP9
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias CLR19.1, NACHT, leucine rich repeat and PYD containing 9, NACHT, LRR and PYD containing protein 9, NACHT, LRR and PYD domains-containing protein 9, NALP9, NLR family, pyrin domain containing 9, NOD6, nucleotide-binding oligomerization domain protein 6, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 9, PAN12, PYRIN and NACHT-containing protein 12
Host Species Rabbit
Immunogen NLRP9 antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human NLRP9. The immunogen is located within amino acids 190 - 240 of NLRP9. Amino Acid Squence: SRDWPESSEKIEDI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 338321
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.