Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 50556660 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-556-660 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-556-660 Supplier Proteintech Group Inc Supplier No. 172151AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NLR family member X1 (NLRX1) belongs to the NLRP family and contains four LRR (leucine-rich) repeats and one NATCH domain. NLRX1 localizes to the mitochondrial outer membrane and interacts through the mitochondrial antiviral signalling adaptor MAVS during viral infection. Expression of NLRX1 results in the potent inhibition of RLH- and MAVS-mediated interferon-beta promoter activity and in the disruption of virus-induced RLH-MAVS interactions. It is also reported that NLRX1 synergistically potentiated ROS production induced by tumour necrosis factor alpha, Shigella infection and double-stranded RNA, resulting in amplified NF-kappaB-dependent and JUN amino-terminal kinases-dependent signalling, thus contributing to the link between ROS generation at the mitochondria and innate immune responses.

Specifications

Antigen NLRX1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NLRX1
Gene Accession No. Q3TL44, Q86UT6
Gene Alias Caterpiller protein 11.3, CLR11.3, DLNB26, NLR family member X1, NLRX1, NOD26, NOD5, NOD9
Gene Symbols NLRX1
Host Species Rabbit
Immunogen NLRX1 Fusion Protein Ag11000
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270151, 79671
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.