Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NM23-H1 Antibody (5F4), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB162616 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162616 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162616 Supplier Novus Biologicals Supplier No. NBP259498R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NM23-H1 Monoclonal antibody specifically detects NM23-H1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen NM23-H1
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 5F4
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.4.6, GAAD, Granzyme A-activated DNase, Metastasis inhibition factor nm23, NB, NBS, NDK A, NDKA, NDP kinase A, NDPKA, NDPK-A, NM23AWD, nm23-H1, non-metastatic cells 1, protein (NM23A) expressed in, nucleoside diphosphate kinase A, Tumor metastatic process-associated protein
Host Species Mouse
Immunogen Recombinant human NM23-H1 (1-152aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4830
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.