Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NM23-H1 Rabbit anti-Human, Clone: 84, Novus Biologicals™
SDP

Catalog No. NB021327 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021327 50 μg
NB021326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021327 Supplier Novus Biologicals Supplier No. NBP28993050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NM23-H1 Monoclonal specifically detects NM23-H1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NM23-H1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 84
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Formulation 0.2 um filtered solution in PBS
Gene Alias EC 2.7.4.6, GAAD, Granzyme A-activated DNase, Metastasis inhibition factor nm23, NB, NBS, NDK A, NDKA, NDP kinase A, NDPKA, NDPK-A, NM23AWD, nm23-H1, non-metastatic cells 1, protein (NM23A) expressed in, nucleoside diphosphate kinase A, Tumor metastatic process-associated protein
Host Species Rabbit
Immunogen Recombinant Human NM23-H1 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4830
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.