Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NM23-H2/NME2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219552 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219552 Supplier Novus Biologicals Supplier No. NBP219552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NM23-H2/NME2 Polyclonal specifically detects NM23-H2/NME2 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen NM23-H2/NME2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal
Gene Alias EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in
Gene Symbols NME2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human nm23-H2. The exact sequence is proprietary.
Molecular Weight of Antigen 17 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4831
Test Specificity Human, Mouse. Species reactivity based on sequence identity: Rabbit (100%), Rat (100%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.