Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NMB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121741
This item is not returnable.
View return policy
Description
NMB stimulates smooth muscle contraction in a manner similar to that of bombesin.
Specifications
NMB | |
Polyclonal | |
Unconjugated | |
NMB | |
3110023K12Rik; neuromedin B; neuromedin-B; Neuromedin-B-32; neuromedin-beta; NMB; RGD1562710 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4828, 68039 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P08949, Q9CR53 | |
NMB | |
Ala25-Lys121 (Accession P08949), with N-terminal His and GST Tag. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction