Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
Antigen | NMB |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121742
|
Invitrogen™
PA5121742 |
20 μL |
Each for $178.40
|
|
|||||
PIPA5121741
|
Invitrogen™
PA5121741 |
100 μL |
Each for $511.50
|
|
|||||
Description
NMB stimulates smooth muscle contraction in a manner similar to that of bombesin.Specifications
NMB | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P08949, Q9CR53 | |
4828, 68039 | |
Ala25-Lys121 (Accession P08949), with N-terminal His and GST Tag. | |
Primary | |
-20°C | |
NMB |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
3110023K12Rik; neuromedin B; neuromedin-B; Neuromedin-B-32; neuromedin-beta; NMB; RGD1562710 | |
NMB | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title