Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDA R, NR1 Subunit Antibody (1031915), PerCP, Novus Biologicals™
SDP

Catalog No. NB305551 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305551 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305551 Supplier Novus Biologicals Supplier No. FAB10655C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

NMDA R, NR1 Subunit Monoclonal specifically detects NMDA R, NR1 Subunit in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry.

Specifications

Antigen NMDA R, NR1 Subunit
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry
Classification Monoclonal
Clone 1031915
Conjugate PerCP
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation PBS
Gene Alias GluN1, MRD8, NDHMSD, NDHMSR,NMD-R1, NMDA1, NMDAR1, NR1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GRIN1/NMDAR1, Met1-Gln559
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2902
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.