Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDA R, NR1 Subunit Antibody, Novus Biologicals™
SDP

Catalog No. NB300114 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300114 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300114 Supplier Novus Biologicals Supplier No. NB300114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NMDA R, NR1 Subunit Polyclonal specifically detects NMDA R, NR1 Subunit in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen NMDA receptor N1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:1000-1:2000
Formulation No Preservative
Gene Alias GluN1, MRD8, NDHMSD, NMDA1, NMDAR1, NR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues specific to the NR1 subunit, N1 splice variant insert conjugated to KLH. Accession # P35439
Molecular Weight of Antigen 120 kDa
Purification Method Antigen Affinity-purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Test Specificity Specific for the ∼120k NR1 subunit, N1 splice variant insert protein of the NMDA receptor. Does not recognize NR1 subunits of the NMDA receptor that do not contain the N1 insert.
Reconstitution Reconstitute with 50 ul PBS to desired concetration.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.