Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDA R, NR1 Subunit Antibody - Splice Variant, Novus Biologicals™
SDP

Catalog No. NB300116 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300116 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300116 Supplier Novus Biologicals Supplier No. NB300116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NMDA R, NR1 Subunit Polyclonal specifically detects NMDA R, NR1 Subunit in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen NMDAR2C Splice Variant
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:1000-1:2000
Formulation No Preservative
Gene Accession No. P35439
Gene Alias GluN1, MRD8, NDHMSD, NMDA1, NMDAR1, NR1
Gene Symbols GRIN2C
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues specific to the NR1 subunit, C2 splice variant insert conjugated to KLH. Accession # P35439
Molecular Weight of Antigen 120 kDa
Purification Method Antigen Affinity-purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2905
Test Specificity Specific for the ∼120k NR1 subunit of the NMDA Receptor containing the C2 splice variant insert. Does not recognize the NR1 subunits of the NMDA receptor that do not contain the C2 insert.
Reconstitution Reconstitute with 50 ul PBS to desired concetration.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.