Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GRIN1/NMDAR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | NMDA R, NR1 Subunit |
---|---|
Clone | 1031915 |
Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, Immunocytochemistry 8-25 ug/mL |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10655SP
![]() |
R&D Systems
MAB10655SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10655100
![]() |
R&D Systems
MAB10655100 |
100 μg |
Each for $432.50
|
|
|||||
Description
NMDA R, NR1 Subunit Monoclonal antibody specifically detects NMDA R, NR1 Subunit in Mouse samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
NMDA R, NR1 Subunit | |
Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
2902 | |
IgG2a | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
1031915 | |
Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Rat | |
Mouse | |
GluN1, MRD8, NDHMSD, NMDA R, NR1 Subunit, NMDA1, NMDAR1, NMD-R1, NR1 | |
Mouse myeloma cell line NS0-derived recombinant mouse GRIN1/NMDAR1, Met1-Gln559 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title