Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NMDA

Click to view available options
Quantity:
50 mg
500 mg
Description
Prototypic NMDA receptor agonist.
Caged version also available (Cat. No 2224).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 147.13 |
| Purity | >99% |
| Molecular Weight (g/mol) | 147.13 |
| Chemical Name or Material | NMDA |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction