Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR1 Antibody (R1JHL), Novus Biologicals™
SDP

Catalog No. NB300118 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.015 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300118 0.015 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300118 Supplier Novus Biologicals Supplier No. NB300118
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 13 publications

NMDAR1 Monoclonal specifically detects NMDAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen NMDAR1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone R1JHL
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 3ug/200ug lysate, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1 ug per ml
Gene Accession No. P35439
Gene Alias glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1
Gene Symbols GRIN1
Host Species Mouse
Immunogen Fusion protein containing amino acids 1-564 of the NMDAR1 subunit. Accession # P35439
Molecular Weight of Antigen 120 kDa
Purification Method Tissue culture supernatant
Quantity 0.015 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2902
Test Specificity Specific for the ∼120k NR1 subunit of the NMDA receptor.
Reconstitution Reconstitute with 50 ul PBS to desired concentration.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.