Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR1, p Ser896 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100820920.025ML
Description
NMDAR1 Polyclonal specifically detects NMDAR1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
NMDAR1 (p Ser896) | |
Polyclonal | |
Unconjugated | |
PBS [pH 7.4] containing 150mM NaCl and 50% glycerol (without Mg2+ and Ca2+) with 0.02% Sodium Azide | |
glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1 | |
Rabbit | |
120 kDa | |
0.025 mL | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
2902 | |
Rat, Mouse, Human | |
IgG |
Western Blot | |
1.0 mg/mL | |
Western Blot 1:500-1:1000 | |
Q05586 | |
GRIN1 | |
The antiserum was produced against synthesized phosphopeptide derived from human NMDAR1 around the phosphorylation site of serine 896 (R-R-SP-S-K). | |
Affinity Purified | |
RUO | |
Primary | |
NMDAR1 (specific to Phospho-Ser896) detects endogenous levels of NMDAR1 only when phosphorylated at serine 896. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction