Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR1, p Ser896 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$243.50 - $402.00

Specifications

Antigen NMDAR1 (p Ser896)
Concentration 1.0 mg/mL
Dilution Western Blot 1:500-1:1000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017868
SDP
View Documents
Novus Biologicals
NB100820920.025ML
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NB10082092
SDP
View Documents
Novus Biologicals
NB10082092
0.05 mL
Each for $402.00
Only null left
Add to Cart
 
Description

Description

NMDAR1 Polyclonal specifically detects NMDAR1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

NMDAR1 (p Ser896)
Western Blot 1:500-1:1000
Polyclonal
Rabbit
Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Q05586
2902
The antiserum was produced against synthesized phosphopeptide derived from human NMDAR1 around the phosphorylation site of serine 896 (R-R-SP-S-K).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot
Unconjugated
RUO
Rat, Mouse, Human
glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1
GRIN1
IgG
Affinity Purified
120 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.