Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2A Antibody, Novus Biologicals™
SDP

Catalog No. NB300105100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300105100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300105100 Supplier Novus Biologicals Supplier No. NB300105100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 9 publications

NMDAR2A Polyclonal specifically detects NMDAR2A in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen NMDAR2A
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:1000 - 1:2000, Immunoprecipitation 3 ul per 200 ug lysate, Immunohistochemistry-Paraffin 1:1000- 1:2000
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. Q00959
Gene Alias glutamate receptor, ionotropic, N-methyl D-aspartate 2A, hNR2A, NMDA receptor subtype 2A, N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor subunit 2A, subunit epsilon-1
Gene Symbols GRIN2A
Host Species Rabbit
Immunogen Fusion protein from the C-terminal region of the NR2A subunit
Molecular Weight of Antigen 180 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroendocrinology, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2903
Test Specificity Specific for the ∼180k NR2A subunit of the NMDA receptor. Recognizes human, mouse and rat forms of the NR2A subunit of NMDAR. No reactivity towards the NR2B and NR2C subunits. Immunolabeling is blocked by pre-adsorption of antibody with the fusion protein used to generate the antibody.
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.