Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2A Antibody (S327-95), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB222405405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222405405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222405405 Supplier Novus Biologicals Supplier No. NBP222405AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NMDAR2A Monoclonal specifically detects NMDAR2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NMDAR2A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S327-95
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GluN2A, glutamate receptor, ionotropic, N-methyl D-aspartate 2A, hNR2A, NMDA receptor subtype 2A, N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor subunit 2A, subunit epsilon-1
Gene Symbols GRIN2A
Host Species Mouse
Immunogen Fusion protein amino acids 75-325 (extracellular N-terminus) of rat GluN2A/NR2A
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Neuroendocrinology, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2903
Test Specificity Detects approx 170kDa. Does not react with NR2B.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.