Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2A Antibody (T7) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019490 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019490 0.025 mg
NBP229863 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019490 Supplier Novus Biologicals Supplier No. NBP2298630.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NMDAR2A Monoclonal specifically detects NMDAR2A in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NMDAR2A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone T7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/ml, Immunohistochemistry 25-5 ug/ml, Immunocytochemistry/Immunofluorescence 25-50ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS with No Preservative
Gene Accession No. Q12879
Gene Alias glutamate receptor, ionotropic, N-methyl D-aspartate 2A, hNR2A, NMDA receptor subtype 2A, N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor subunit 2A, subunit epsilon-1
Gene Symbols GRIN2A
Host Species Mouse
Immunogen Amino acid region 1099-1213 from human NMDAR2A produced as a GST fusion protein.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2903
Test Specificity Amino acid region 1099-1213 from human NMDAR2A produced as a GST fusion protein.
Target Species Rat, Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.