Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR2A Antibody (T7) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $890.00
Specifications
| Antigen | NMDAR2A |
|---|---|
| Clone | T7 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-5 ug/ml, Immunohistochemistry 25-5 ug/ml, Immunocytochemistry/Immunofluorescence 25-50ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunohistochemistry |
Description
NMDAR2A Monoclonal specifically detects NMDAR2A in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NMDAR2A | |
| 1.0 mg/mL | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Rat, Human | |
| 2903 | |
| Amino acid region 1099-1213 from human NMDAR2A produced as a GST fusion protein. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| T7 | |
| Western Blot 1-5 ug/ml, Immunohistochemistry 25-5 ug/ml, Immunocytochemistry/Immunofluorescence 25-50ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| Q12879 | |
| GRIN2A | |
| IgG1 | |
| Affinity Purified | |
| Amino acid region 1099-1213 from human NMDAR2A produced as a GST fusion protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title