Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR2B Antibody (BWJHL), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP229790
Description
NMDAR2B Monoclonal specifically detects NMDAR2B in Human, Rat samples. It is validated for Western Blot, Simple Western.Specifications
| NMDAR2B | |
| Monoclonal | |
| Unconjugated | |
| No buffer with No Preservative | |
| glutamate [NMDA] receptor subunit epsilon-2, glutamate receptor subunit epsilon-2, glutamate receptor, ionotropic, N-methyl D-aspartate 2B, hNR3, MGC142180, NMDAR2BMGC142178, N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 3, NR2B, NR3 | |
| Mouse | |
| 180 kDa | |
| 0.1 mg | |
| Autophagy, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 2904 | |
| Human, Rat | |
| IgG |
| Western Blot | |
| BWJHL | |
| Western Blot 1:100-1:2000 | |
| Q13224 | |
| GRIN2B | |
| GST fusion protein corresponding to residues 934-1457 of rat NR2B. Clone BWJHL. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NR2B. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction