Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR2B Antibody (BWJHL), Novus Biologicals™

Mouse Monoclonal Antibody
$854.00
Specifications
| Antigen | NMDAR2B |
|---|---|
| Clone | BWJHL |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
NMDAR2B Monoclonal specifically detects NMDAR2B in Human, Rat samples. It is validated for Western Blot, Simple Western.Specifications
| NMDAR2B | |
| Western Blot 1:100-1:2000 | |
| Monoclonal | |
| Mouse | |
| Autophagy, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| No buffer with No Preservative | |
| glutamate [NMDA] receptor subunit epsilon-2, glutamate receptor subunit epsilon-2, glutamate receptor, ionotropic, N-methyl D-aspartate 2B, hNR3, MGC142180, NMDAR2BMGC142178, N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 3, NR2B, NR3 | |
| GRIN2B | |
| IgG | |
| Affinity Purified | |
| NR2B. |
| BWJHL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| Q13224 | |
| 2904 | |
| GST fusion protein corresponding to residues 934-1457 of rat NR2B. Clone BWJHL. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 180 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title