Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2B, p Tyr1252 Antibody, Novus Biologicals™
SDP

Catalog No. NB300292 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300292 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300292 Supplier Novus Biologicals Supplier No. NB300292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NMDAR2B Polyclonal specifically detects NMDAR2B in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NMDAR2B (p Tyr1252)
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:400
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. Q00960
Gene Alias glutamate [NMDA] receptor subunit epsilon-2, glutamate receptor subunit epsilon-2, glutamate receptor, ionotropic, N-methyl D-aspartate 2B, hNR3, MGC142180, NMDAR2BMGC142178, N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 3, NR2B, NR3
Gene Symbols GRIN2B
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr1252 conjugated to KLH. Accession # Q00960
Molecular Weight of Antigen 180 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2904
Test Specificity Specific for ∼180k NMDAR NR2B subunit protein phosphorylated at Tyr1252. Immunolabeling of the NMDA NR2B subunit band is blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide. Immunolabeling is also blocked by lambda-phosphatase treatment. The antibody may also show some slight reactivity with Tyr1246 of NR2A.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.