Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2C Antibody, Novus Biologicals™
SDP

Catalog No. NB300107 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.01 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300107 0.01 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300107 Supplier Novus Biologicals Supplier No. NB300107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

NMDAR2C Polyclonal specifically detects NMDAR2C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen NMDA R, NR2C Subunit
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 3 ul per 200 ug lysate
Formulation No Preservative
Gene Accession No. Q00961
Gene Alias glutamate [NMDA] receptor subunit epsilon-3, glutamate receptor, ionotropic, N-methyl D-aspartate 2C, NMDAR2CNR2C, N-methyl D-aspartate receptor subtype 2C, N-methyl-D-aspartate receptor subunit 2C
Gene Symbols GRIN2C
Host Species Rabbit
Immunogen Fusion protein from the N-terminal region of the NMDAR2C subunit. Accession # Q00961
Molecular Weight of Antigen 140 kDa
Purification Method Antigen Affinity-purified
Quantity 0.01 mg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2905
Test Specificity Specific for the ∼140k NR2C subunit of the NMDA receptor. Also labels the ∼180k NR2A and the ∼180k NR2B subunits of the NMDA receptor. Immunolabeling is blocked by preadsorption of antibody with the immunogen that was used to generate the antibody.
Reconstitution Reconstitute with 50 ul PBS to desired concetration.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.