Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR2C Mouse anti-Rat, Unconjugated, Clone: N422/18, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | NMDAR2C |
---|---|
Clone | N422/18 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN18692MI
|
MilliporeSigma
MABN186925UG |
25 μg |
Each for $182.31
|
|
|||||
MABN18691MI
|
MilliporeSigma
MABN1869100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
NMDAR2C Monoclonal antibody specifically detects NMDAR2C in Rat samples. It is validated for Western Blotting.Specifications
NMDAR2C | |
Western Blot | |
Unconjugated | |
Mouse | |
Rat | |
Glutamate receptor ionotropic+C6 NMDA 2C;GluN2C;Glutamate [NMDA] receptor subunit epsilon-3;N-methyl D-aspartate receptor subtype 2C;NR2C | |
GST-tagged recombinant fragment corresponding to 398 amino acids from the C-terminal, cytoplasmic domain of rat Glutamate receptor ionotropic, NMDAR2C. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼150 kDa observed; 135.27 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
N422/18 | |
Monoclonal | |
Purified | |
Neuroscience | |
NP_036707.3 | |
Grin2c | |
IgG1 κ | |
Protein G Purified | |
Clone N422/18 is a mouse monoclonal antibody that specifically detects rat Glutamate receptor ionotropic, NMDAR2C. It targets an epitope within the C-terminal cytoplasmic domain. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title