Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR3B Antibody (2A8E11) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261747 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61747 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61747 Supplier Novus Biologicals Supplier No. NBP261747
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NMDAR3B Monoclonal specifically detects NMDAR3B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen NMDAR3B
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 2A8E11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias glutamate [NMDA] receptor subunit 3B, glutamate receptor, ionotropic, N-methyl-D-aspartate 3B, NMDA receptor subunit 3B, NMDA type glutamate receptor subunit NR3B, NMDAR3B, N-methyl-D-aspartate receptor subtype 3B, NR3B
Gene Symbols GRIN3B
Host Species Mouse
Immunogen Purified recombinant fragment of human GRIN3B (AA: 135-276) expressed in E. Coli.
Molecular Weight of Antigen 113 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 116444
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.