Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR3B Antibody (2A8E11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | NMDAR3B |
---|---|
Clone | 2A8E11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61747
![]() |
Novus Biologicals
NBP261747 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
NMDAR3B Monoclonal specifically detects NMDAR3B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
NMDAR3B | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
GRIN3B | |
IgG2a | |
Protein G purified |
2A8E11 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
116444 | |
Purified recombinant fragment of human GRIN3B (AA: 135-276) expressed in E. Coli. | |
Primary | |
113 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title