Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR3B Antibody, Novus Biologicals™
SDP

Catalog No. NBP152062 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152062 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152062 Supplier Novus Biologicals Supplier No. NBP152062
Add to Cart
Add to Cart

Goat Polyclonal Antibody

NMDAR3B Polyclonal specifically detects NMDAR3B in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen NMDAR3B
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_619635.1
Gene Alias glutamate [NMDA] receptor subunit 3B, glutamate receptor, ionotropic, N-methyl-D-aspartate 3B, NMDA receptor subunit 3B, NMDA type glutamate receptor subunit NR3B, NMDAR3B, N-methyl-D-aspartate receptor subtype 3B, NR3B
Gene Symbols GRIN3B
Host Species Goat
Immunogen Peptide with sequence C-EAPPHSGRPGSQ corresponding to C-Terminus according to NP_619635.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116444
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.