Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDARA1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB341657 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341657 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341657 Supplier Novus Biologicals Supplier No. NBP281973MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NMDARA1 Polyclonal antibody specifically detects NMDARA1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen NMDARA1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias glutamate [NMDA] receptor-associated protein 1, glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1(glutamate binding), glutamate receptor, NMDA subtype, glutamate-binding subunit, HNRGW, LFG1, NMDA receptor glutamate-binding subunit, NMDARA1MGC99687, Putative MAPK-activating protein PM02, TMBIM3, transmembrane BAX inhibitor motif containing 3
Host Species Rabbit
Immunogen Rabbit polyclonal NMDARA1 antibody was raised against an 18 amino acid peptide near the center of human NMDARA1. The immunogen is located within amino acids 110 - 160 of NMDARA1. Amino Acid Squence: QDPDSPQHGNYQEEGPPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2907
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.