Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NME3 Polyclonal Antibody

Rabbit Polyclonal Antibody

$209.50 - $511.50

Specifications

Antigen NME3
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119311
View Documents
Invitrogen™
PA5119311
200 μL
Each for $511.50
Only null left
Add to Cart
 
PIPA5121740
View Documents
Invitrogen™
PA5121740
20 μL
Each for $209.50
Only null left
Add to Cart
 
Description

Description

NME3 Polyclonal Antibody for Western Blot, IHC (P)

NME3 mRNA is preferentially expressed at early stages of myeloid differentiation of highly purified CD34(+) cells. Its constitutive expression in a myeloid precursor line, which is growth-factor dependent for both proliferation and differentiation, results in inhibition of granulocytic differentiation induced by granulocyte colony-stimulating factor and causes apoptotic cell death. These results appear consistent with a role for the NME3 gene in normal hematopoiesis and raise the possibility that its overexpression contributes to differentiation arrest, a feature of blastic transformation in chronic myelogenous leukemia.
Specifications

Specifications

NME3
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Mouse, Pig
NME3
1810009F08Rik; AI413736; c371H6.2; DR-nm23; expressed in non-metastatic cells 3; expressed in non-metastatic cells 3 protein (nucleoside diphosphate kinase); expressed in non-metastatic cells 3, protein (nucleoside diphosphate kinase); NDK 3; Ndk3; NDP kinase 3; NDP kinase C; NDPKC; NDPK-C; NM23-dr; NM23H3; nm23-H3; nm23-M3; NME/NM23 nucleoside diphosphate kinase 3; Nme3; non-metastatic cell expressed protein 3; non-metastatic cells 3, protein expressed in; nucleoside diphosphate kinase 3; Nucleoside diphosphate kinase C; nucleoside diphosphate kinase DR-nm23
NME3
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q13232, Q9WV85
100514398, 4832, 79059
Recombinant protein Non Metastatic Cells 3, Protein NM23A. The antigen corresponds to amino acid range 1-168 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.