Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NME4 Antibody (OTI1A5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP27305A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27305A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27305A594 Supplier Novus Biologicals Supplier No. NBP273005AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NME4 Monoclonal specifically detects NME4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NME4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A5
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 2.7.4.6, NDP kinase, mitochondrial, NDPK-D, NM23D, nm23-H4mitochondrial, non-metastatic cells 4, protein expressed in
Gene Symbols NME4
Host Species Mouse
Immunogen Full length recombinant protein of human NME4 (NP_005000) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4833.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.