Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NME4 Antibody (OTI1A5), Novus Biologicals™
SDP

Catalog No. NBP202487 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP202487 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP202487 Supplier Novus Biologicals Supplier No. NBP202487
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NME4 Monoclonal specifically detects NME4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NME4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1A5
Concentration 0.59 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50
Gene Alias EC 2.7.4.6, NDP kinase, mitochondrial, NDPK-D, NM23D, nm23-H4mitochondrial, non-metastatic cells 4, protein expressed in
Gene Symbols NME4
Host Species Mouse
Immunogen Full length human recombinant protein of human NME4 (NP_005000) produced in HEK293T cell.
Molecular Weight of Antigen 20.5 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4833
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.