Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMRAL1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB326169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326169 Supplier Novus Biologicals Supplier No. NBP299301JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NMRAL1 Polyclonal antibody specifically detects NMRAL1 in Human samples. It is validated for Western Blot

Specifications

Antigen NMRAL1
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias HSCARGFLJ25918, NmrA-like family domain containing 1, nmrA-like family domain-containing protein 1, SDR48A1, short chain dehydrogenase/reductase family 48A, member 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NMRAL1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57407
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.