Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMT2 Monoclonal Antibody (OTI2B4), TrueMAB™, OriGene
SDP

Catalog No. 502540466
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0466 30 μL
50-167-4922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0466 Supplier OriGene Supplier No. TA504176S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NMT2 Monoclonal antibody specifically detects NMT2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen NMT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B4
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene JCHAIN
Gene Accession No. P01591
Gene Alias A930001K02Rik; AI605445; AU044698; fb40e03; Glycylpeptide N-tetradecanoyltransferase 2; glycylpeptide N-tetradecanoyltransferase 2 variant 3; glycylpeptide N-tetradecanoyltransferase 2 variant 4; hNMT-2; myristoyl-CoA:protein N-myristoyltransferase 2; NMT 2; Nmt2; NMT2 protein; N-myristoyltransferase 2; peptide N-myristoyltransferase 2; Type II N-myristoyltransferase; wu:fb40e03
Host Species Mouse
Immunogen Full length human recombinant protein of human IGJ produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3512
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.