Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NNC 0640

Click to view available options
Quantity:
10 mg
50 mg
Description
NNC 0640 is a negative allosteric modulator of glucagon receptors (pKi = 7.4). Binds to external surface of the transmembrane domain. Also negative allosteric modulator of glucagon-like peptide 1 (GLP-1) receptor. Inhibits GLP-1-mediated cAMP accumulation in vitro.
Specifications
Specifications
Chemical Name or Material | 4-[[(4-Cyclohexylphenyl)[[[3-(methylsulfonyl)phenyl]amino]carbonyl]amino]methyl]-N-2 H-tetrazol-5-ylbenzamide |
CAS | 307986-98-7 |
Quantity | 10 mg |
Target | Glucagon-Like Peptide 1 Receptor Modulators |
Molecular Formula | C29H31N7O4S |
Formula Weight | 2172 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction