Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NNC 711
GREENER_CHOICE

Catalog No. 177910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-791-0 10 mg
17-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-791-0 Supplier Tocris Bioscience™ Supplier No. 1779/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective GAT-1 inhibitor

Potent and selective inhibitor of GABA uptake by GAT-1 (IC50 values are 0.04, 171, 1700 and 622 μM for hGAT-1, rGAT-2, hGAT-3 and hBGT-1 respectively). Anticonvulsant following systemic administration in vivo. Also exhibits cognition-enhancing activity.

Chemical Identifiers

CAS 145645-62-1
Molecular Formula C21H23ClN2O3

Specifications

Quantity 10 mg
Formula Weight 386.88
Purity >99%
Molecular Weight (g/mol) 386.876
Chemical Name or Material NNC 711
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.